Dimethyl oxalate, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
M-Clarity™ quality level = MQ100
135623-100G
135623-500G
135623-5G
SIAL135623-100GEA
47.6
EUR
SIAL135623-100G
SIAL135623-500G
SIAL135623-5G
Dimethyl oxalate, Sigma-Aldrich®
Dimethyl oxalate
Formula:
C₄H₆O₄ Boiling Pt: 163 °C (1013 hPa) Melting Pt: 54 °C Density: 1.422 g/cm³ (15 °C) Flash Pt: 62 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008442 CAS Number: 553-90-2 EINECS: 209-053-6 UN: 3261 ADR: 8,III Merck Index: 12,06181 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...