Triethyl citrate, Sigma-Aldrich®
Supplier: Merck
Synonyms: Triethyl 2-hydroxypropane-1,2,3-tricarboxylate, Hydroxytricarballylic acid triethyl ester, Citric acid triethyl ester
M-Clarity™ quality level = MQ300
W308307-10KG-K W308307-1KG-K W308307-25KG-K
SIALW308307-10KG-KEA 506 EUR
SIALW308307-10KG-K SIALW308307-1KG-K SIALW308307-25KG-K
Triethyl citrate, Sigma-Aldrich®
Triethyl citrate
Formula: C₁₂H₂₀O₇
MW: 276.29 g/mol
Boiling Pt: 294
Melting Pt: –55 °C
Density: 1.135 g/cm³ (25 °C)
Flash Pt: 150
MDL Number: MFCD00009201
CAS Number: 77-93-0
EINECS: 201-070-7