Butyric acid, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Butanoic acid
M-Clarity™ quality level = MQ200
B103500-250ML B103500-5ML B103500-100ML B103500-1L B103500-500ML
SIALB103500-250MLEA 42.2 EUR
SIALB103500-250ML SIALB103500-5ML SIALB103500-100ML SIALB103500-1L SIALB103500-500ML
Butyric acid, Sigma-Aldrich®
Butyric acid
Formula: C₄H₈O₂
MW: 88.11 g/mol
Boiling Pt: 163.5
Melting Pt: –3 °C
Density: 0.958 g/cm³ (20 °C)
Flash Pt: 67
Storage Temperature: Ambient
MDL Number: MFCD00002814
CAS Number: 107-92-6
EINECS: 203-532-3
UN: 2820
ADR: 8 III
Merck Index: 13,01593