Lauric acid, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
Synonyms:
Dodecanoic acid, Dodecylic acid
M-Clarity™ quality level = MQ200
L556-1KG
L556-25G
L556-3KG
SIALL556-1KGEA
52.2
EUR
SIALL556-1KG
SIALL556-25G
SIALL556-3KG
Lauric acid, Sigma-Aldrich®
Lauric acid
Formula:
C₁₂H₂₄O₂ Boiling Pt: 296 °C (1013 hPa) Melting Pt: 42…45 °C Density: 0.875 g/cm³ (20 °C) Flash Pt: >150 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002736 CAS Number: 143-07-7 EINECS: 205-582-1 Merck Index: 13,05400 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...