Lauric acid, Sigma-Aldrich®

Supplier: Merck
Warning


M-Clarity™ quality level = MQ300

W261416-10KG-K W261416-1KG-K W261416-5KG-K
SIALW261416-10KG-KEA 525 EUR
SIALW261416-10KG-K SIALW261416-1KG-K SIALW261416-5KG-K
Lauric acid, Sigma-Aldrich®
Lauric acid
Formula: C₁₂H₂₄O₂
Boiling Pt: 296 °C (1013 hPa)
Melting Pt: 42…45 °C
Density: 0.875 g/cm³ (20 °C)
Flash Pt: >150 °C
Storage Temperature: Ambient
MDL Number: MFCD00002736
CAS Number: 143-07-7
EINECS: 205-582-1
Merck Index: 13,05400

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR