Palmitic acid, Sigma-Aldrich®
Supplier: Merck
Synonyms: Hexadecanoic acid, Hexadecylic acid
M-Clarity™ quality level = MQ200
P5585-100G P5585-10G P5585-25G
SIALP5585-100GEA 385 EUR
SIALP5585-100G SIALP5585-10G SIALP5585-25G
Palmitic acid, Sigma-Aldrich®
Palmitic acid
Formula: C₁₆H₃₂O₂
MW: 256.43 g/mol
Boiling Pt: 352
Melting Pt: 61…63 °C
Density: 0.853 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002747
CAS Number: 57-10-3
EINECS: 200-312-9
Merck Index: 13,07064