Palmitic acid, Sigma-Aldrich®
Supplier: Merck
Synonyms: Hexadecanoic acid, Hexadecylic acid
M-Clarity™ quality level = MQ300
W283207-10KG-K W283207-1KG-K W283207-25KG-K
SIALW283207-10KG-KEA 216 EUR
SIALW283207-10KG-K SIALW283207-1KG-K SIALW283207-25KG-K
Palmitic acid, Sigma-Aldrich®
Palmitic acid
Formula: C₁₆H₃₂O₂
Boiling Pt: 352 °C (1013 hPa)
Melting Pt: 61…63 °C
Density: 0.853 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002747
CAS Number: 57-10-3
EINECS: 200-312-9
Merck Index: 13,07064