Caprylic acid, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ100
C2875-10ML C2875-100ML C2875-1L C2875-500ML
SIALC2875-10MLEA 38.9 EUR
SIALC2875-10ML SIALC2875-100ML SIALC2875-1L SIALC2875-500ML
Caprylic acid, Sigma-Aldrich®
Caprylic acid
Formula: C₈H₁₆O₂
Boiling Pt: 239.7 °C (1013 hPa)
Melting Pt: 16 °C
Density: 0.9066 g/cm³ (20 °C)
Flash Pt: 130 °C
Storage Temperature: Ambient
MDL Number: MFCD00004429
CAS Number: 124-07-2
EINECS: 204-677-5
UN: 3265
ADR: 8,III
Merck Index: 13,01771