Cesium acetate, Sigma-Aldrich®

Supplier: Merck

Synonyms: Acetic acid cesium salt

M-Clarity™ quality level = MQ100

329827-100G 329827-25G
SIAL329827-100GEA 316 EUR
SIAL329827-100G SIAL329827-25G
Cesium acetate, Sigma-Aldrich®
Cesium acetate
Formula: H₃CCOOCs
MW: 191.95 g/mol
Melting Pt: 194
MDL Number: MFCD00013056
CAS Number: 3396-11-0
EINECS: 222-248-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR