Sodium acetate, anhydrous ≥99.0% ACS
Supplier: Thermo Fisher Scientific
Warning
|
Synonyms:
Acetic acid sodium salt
11554.30
11554.A1
11554.A7
11554.30EA
42.6
EUR
11554.30
11554.A1
11554.A7
Sodium acetate, anhydrous ≥99.0% ACS
Sodium acetate
Purity: 99.0% min
CAS No.: 127-09-3
Molecular Formula: NaOOCCH3
Formula Weight: 82.03
Physical Form: Crystalline
Density: 1.528
pH: 7,5-9
Melting Point: 324°
Beilstein Reference: 3595639
Merck Reference: 14,8571
MDL No.: MFCD00012459
Formula:
H₃CCOONa MW: 82.03 g/mol Boiling Pt: 881.4 Melting Pt: 324 Density: 1.528 Flash Pt: >250 °C (closed cup) Storage Temperature: Ambient |
MDL Number:
MFCD00012459 CAS Number: 127-09-3 EINECS: 204-823-8 REACH: 01-2119485123-42 Merck Index: 14,8571 |