Sodium acetate
Supplier: Roth Carl
X891.1
X891.2
X891.3
X891.5
ROTHX891.1EA
23.2
EUR
ROTHX891.1
ROTHX891.2
ROTHX891.3
ROTHX891.5
Sodium acetate
Sodium acetate
Formula:
H₃CCOONa Boiling Pt: 881.4 °C (1013 hPa) Melting Pt: 324 °C Density: 1.528 g/cm³ (20 °C) Flash Pt: >250 °C (closed cup) Storage Temperature: Ambient |
MDL Number:
MFCD00012459 CAS Number: 127-09-3 EINECS: 204-823-8 REACH: 01-2119485123-42 Merck Index: 12,08711 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...