Sodium acetate, Sigma-Aldrich®
Supplier: Merck
Synonyms: Acetic acid sodium salt
M-Clarity™ quality level = MQ200
S2889-250G S2889-1KG S2889-5KG
SIALS2889-250GEA 60.7 EUR
SIALS2889-250G SIALS2889-1KG SIALS2889-5KG
Sodium acetate, Sigma-Aldrich®
Sodium acetate
Formula: H₃CCOONa
MW: 82.03 g/mol
Boiling Pt: 881.4
Melting Pt: 324
Density: 1.528 g/cm³ (20 °C)
Flash Pt: >250 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00012459
CAS Number: 127-09-3
EINECS: 204-823-8
REACH: 01-2119485123-42
Merck Index: 12,08711