Sodium acetate, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ300
W302406-10KG-K W302406-1KG-K W302406-25KG-K
SIALW302406-10KG-KEA 223 EUR
SIALW302406-10KG-K SIALW302406-1KG-K SIALW302406-25KG-K
Sodium acetate, Sigma-Aldrich®
Sodium acetate
Formula: H₃CCOONa
Boiling Pt: 881.4 °C (1013 hPa)
Melting Pt: 324 °C
Density: 1.528 g/cm³ (20 °C)
Flash Pt: >250 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00012459
CAS Number: 127-09-3
EINECS: 204-823-8
REACH: 01-2119485123-42
Merck Index: 12,08711