Barium acetate, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Barium di(acetate), Acetic acid barium salt
M-Clarity™ quality level = MQ100
255912-10G 255912-50G
SIAL255912-10GEA 191 EUR
SIAL255912-10G SIAL255912-50G
Barium acetate, Sigma-Aldrich®
Barium acetate
Formula: (H₃CCOO)₂Ba
Melting Pt: ∼450 °C
Density: 2.468 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00012447
CAS Number: 543-80-6
EINECS: 208-849-0
UN: 1564
ADR: 6.1,III
Merck Index: 13,00968