Ammonium acetate, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Acetic acid ammonium salt
M-Clarity™ quality level = MQ100
A7330-100G A7330-2.5KG A7330-500G
SIALA7330-100GEA 31.6 EUR
SIALA7330-100G SIALA7330-2.5KG SIALA7330-500G
Ammonium acetate, Sigma-Aldrich®
Ammonium acetate
Formula: H₃CCOONH₄
Melting Pt: 114 °C
Density: 1.17 g/cm³ (20 °C)
Flash Pt: 136 °C
Storage Temperature: Ambient
MDL Number: MFCD00013066
CAS Number: 631-61-8
EINECS: 211-162-9
Merck Index: 13,00495
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd160345483553318b94053de396335a9b18425f&vid=c6375b539008d6b43f50408d30b61261444faebc&prid=web&clid=SAPDAM