Silver(I) acetate, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ200
204374-10G 204374-50G
SIAL204374-10GEA 168 EUR
SIAL204374-10G SIAL204374-50G
Silver(I) acetate, Sigma-Aldrich®
Silver (I) acetate
Formula: C₂H₃AgO₂
Melting Pt: 2000 °C
Density: 3.26 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00012446
CAS Number: 563-63-3
EINECS: 209-254-9
Merck Index: 12,08648