Zinc stearate, Sigma-Aldrich®
Supplier: Merck
Synonyms: Stearic acid zinc salt, Zinc(II) stearate
M-Clarity™ quality level = MQ200
307564-1KG 307564-25G 307564-3KG
SIAL307564-1KGEA 58.9 EUR
SIAL307564-1KG SIAL307564-25G SIAL307564-3KG
Zinc stearate, Sigma-Aldrich®
Zinc stearate
Formula: C₃₆H₇₀O₄Zn
Melting Pt: 140 °C
Density: 1.095 g/cm³ (20 °C)
Flash Pt: 276 °C
Storage Temperature: Ambient
MDL Number: MFCD00013031
CAS Number: 557-05-1
EINECS: 209-151-9
Merck Index: 13,10212
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f8ad7f3e101681798edbd54e57ad5d2ac82b2e5d&vid=435308cdcb35862fe6cc06384da2ef10d84c5700&prid=web&clid=SAPDAM