Cobalt(II) acetate, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Cobaltous acetate, Cobalt acetate, Acetic acid cobalt (II) salt, Cobalt diacetate
M-Clarity™ quality level = MQ200
399973-10G 399973-1G
SIAL399973-10GEA 238 EUR
SIAL399973-10G SIAL399973-1G
Cobalt(II) acetate, Sigma-Aldrich®
Cobalt (II) acetate
Formula: C₄H₆CoO₄
MW: 177.02 g/mol
Melting Pt: 297…299 °C
Density: 1.7755 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008689
CAS Number: 71-48-7
EINECS: 200-755-8
UN: 3077
ADR: 9 III
Merck Index: 12,02494