12-Aminolauric acid, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
159247-25G
159247-5G
SIAL159247-25GEA
120
EUR
SIAL159247-25G
SIAL159247-5G
12-Aminolauric acid, Sigma-Aldrich®
12-Aminolauric acid
Formula:
C₁₂H₂₅NO₂ Boiling Pt: 343 °C (1013 hPa) Melting Pt: 183 °C |
MDL Number:
MFCD00008153 CAS Number: 693-57-2 EINECS: 211-754-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...