L-Methionine, Sigma-Aldrich®
Supplier: Merck
Synonyms: (S)-(-)-2-Amino-4-(methylthio)butyric acid, (S)-(-)-Methionine, L(-)-Methionine, (S)-(-)-2-Amino-4-(methylthio)butanoic acid, H-Met-OH, Methionine, (S)-2-Amino-4-(methylthio)butanoic acid
M-Clarity™ quality level = MQ200
M9625-100G M9625-1KG M9625-25G M9625-500G M9625-5G
SIALM9625-100GEA 99 EUR
SIALM9625-100G SIALM9625-1KG SIALM9625-25G SIALM9625-500G SIALM9625-5G
L-Methionine, Sigma-Aldrich®
L-Methionine
Formula: C₅H₁₁NO₂S
Boiling Pt: 278 °C (1013 hPa)
Melting Pt: 136 °C
Density: 1.373 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00063097
CAS Number: 63-68-3
EINECS: 200-562-9
Merck Index: 13,06004