Sarcosine, Sigma-Aldrich®

Supplier: Merck

Synonyms: H-Sar-OH, N-Methylglycine, Methylaminoacetic acid

M-Clarity™ quality level = MQ200

S7672-100G S7672-10MG S7672-25G
SIALS7672-100GEA 556 EUR
SIALS7672-100G SIALS7672-10MG SIALS7672-25G
Sarcosine, Sigma-Aldrich®
Sarcosine
Formula: C₃H₇NO₂
Boiling Pt: 185 °C (1013 hPa)
Melting Pt: 63 °C
Density: 1.301 g/cm³ (20 °C)
MDL Number: MFCD00004279
CAS Number: 107-97-1
EINECS: 203-538-6
Merck Index: 12,08519

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR