Glycolic acid, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: 2-Hydroxy acid, 2-Hydroxyacetic acid
M-Clarity™ quality level = MQ200
124737-100G 124737-12KG 124737-25G 124737-500G
SIAL124737-100GEA 77 EUR
SIAL124737-100G SIAL124737-12KG SIAL124737-25G SIAL124737-500G
Glycolic acid, Sigma-Aldrich®
Glycolic acid
Formula: C₂H₄O₃
Boiling Pt: 266 °C (1013 hPa)
Melting Pt: 79…80 °C
Density: 1.49 g/cm³ (20 °C)
Flash Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00004312
CAS Number: 79-14-1
EINECS: 201-180-5
UN: 3261
ADR: 8,II
Merck Index: 13,04511