Mercaptoacetic acid (thioglycolic acid), Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ100
528056-100ML 528056-25ML
SIAL528056-100MLEA 27.9 EUR
SIAL528056-100ML SIAL528056-25ML
Mercaptoacetic acid (thioglycolic acid), Sigma-Aldrich®
Mercaptoacetic acid (Thioglycolic acid)
Formula: HSCH₂COOH
Boiling Pt: 96 °C (1013 hPa)
Melting Pt: –16.5 °C
Density: 1.33 g/cm³ (20 °C)
Flash Pt: 130 °C (closed cup)
Storage Temperature: Refrigerator
MDL Number: MFCD00004876
CAS Number: 68-11-1
EINECS: 200-677-4
UN: 1940
ADR: 8,II
Merck Index: 13,09410
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=45179825145ec44f2e058197a0ee54845ae7c750&vid=51e43f5b69b880b196570b3065f9b4d8db7336d1&prid=web&clid=SAPDAM