Sebacic acid, Sigma-Aldrich®
Supplier: Merck
Synonyms: Decanedioic acid, 1,8-Octanedicarboxylic acid, 1,10-Decanedioic acid
84809-1KG 84809-100G
SIAL84809-1KGEA 98.3 EUR
SIAL84809-1KG SIAL84809-100G
Sebacic acid, Sigma-Aldrich®
Sebacic acid
Formula: C₁₀H₁₈O₄
MW: 202.25 g/mol
Boiling Pt: 374
Melting Pt: 127…131 °C
Density: 1.21 g/cm³ (20 °C)
Flash Pt: 221
Storage Temperature: Ambient
MDL Number: MFCD00004440
CAS Number: 111-20-6
EINECS: 203-845-5
REACH: 01-2119519212-52
Merck Index: 12,08558