Malonic acid ≥99.5%, Reagent Grade
Supplier: Thermo Fisher Scientific
Danger
31715.22 31715.36
31715.22EA 47 EUR
31715.22 31715.36
Malonic acid ≥99.5%, Reagent Grade
Malonic acid
Formula: C₃H₄O₄
Boiling Pt: 387 °C (1013 hPa)
Melting Pt: 132…136 °C (dec.)
Density: 1.62
Flash Pt: 157 °C (314 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002707
CAS Number: 141-82-2
EINECS: 205-503-0
Merck Index: 14,5710