Succinic acid, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
1,4-Butanedioic acid
M-Clarity™ quality level = MQ200
S3674-100G
S3674-1KG
SIALS3674-100GEA
71.7
EUR
SIALS3674-100G
SIALS3674-1KG
Succinic acid, Sigma-Aldrich®
Succinic acid
Formula:
HOOCCH₂CH₂COOH Boiling Pt: 235 °C (1013 hPa) Melting Pt: 182 °C Density: 1.56 g/cm³ (20 °C) Flash Pt: 206 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002789 CAS Number: 110-15-6 EINECS: 203-740-4 UN: 3261 ADR: 8,III Merck Index: 13,08953 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...