Oxalic acid ≥98%, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: Ethanedioic acid

M-Clarity™ quality level = MQ200

194131-1KG 194131-250G 194131-5G
SIAL194131-1KGEA 154 EUR
SIAL194131-1KG SIAL194131-250G SIAL194131-5G
Oxalic acid ≥98%, Sigma-Aldrich®
Oxalic acid
Formula: C₂H₂O₄
Boiling Pt: 365 °C (1013 hPa)
Melting Pt: 99…100 °C
Density: 1.9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002573
CAS Number: 144-62-7
EINECS: 205-634-3
UN: 3261
ADR: 8,III
Merck Index: 13,06980

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR