Oxalic acid ≥99.999% (trace metals basis), purified, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
M-Clarity™ quality level = MQ200
658537-100G
658537-25G
SIAL658537-100GEA
225
EUR
SIAL658537-100G
SIAL658537-25G
Oxalic acid ≥99.999% (trace metals basis), purified, Sigma-Aldrich®
Oxalic acid
Formula:
C₂H₂O₄ Boiling Pt: 365 °C (1013 hPa) Melting Pt: 99…100 °C Density: 1.9 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002573 CAS Number: 144-62-7 EINECS: 205-634-3 UN: 3261 ADR: 8,III Merck Index: 13,06980 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...