Oxalic acid ≥99.0% (by redox titration), puriss. p.a. analytical reagent, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Ethanedioic acid
75688-250G 75688-50G
SIAL75688-250GEA 88.5 EUR
SIAL75688-250G SIAL75688-50G
Oxalic acid ≥99.0% (by redox titration), puriss. p.a. analytical reagent, Sigma-Aldrich®
Oxalic acid
Formula: C₂H₂O₄
MW: 90.04 g/mol
Boiling Pt: 365
Melting Pt: 99…100 °C
Density: 1.9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002573
CAS Number: 144-62-7
EINECS: 205-634-3
UN: 3261
ADR: 8 III
Merck Index: 13,06980