EDTA tetrasodium salt, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
Ethylenediaminetetraacetic acid tetrasodium salt, Ethylenedinitrilotetra-acetic acid tetrasodium salt, Tetrasodium ethylenediaminetetraacetate, EDTA tetrasodium, Edetic acid tetrasodium salt, Kestranal® 4S
M-Clarity™ quality level = MQ200
27261-1KG-R
27261-25KG-R
SIAL27261-1KG-REA
67.1
EUR
SIAL27261-1KG-R
SIAL27261-25KG-R
EDTA tetrasodium salt, Sigma-Aldrich®
EDTA tetrasodium salt
Formula:
C₁₀H₁₂N₂Na₄O₈ Melting Pt: > 300 °C Density: 6.9 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00150027 CAS Number: 64-02-8 EINECS: 200-573-9 Merck Index: 12,03557 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...