Ethyl fluoroacetate, Sigma-Aldrich®

Supplier: Merck

Synonyms: Fluoroacetic ethyl ester

M-Clarity™ quality level = MQ100

163813-10G 163813-50G
SIAL163813-10GEA 49.8 EUR
SIAL163813-10G SIAL163813-50G
Ethyl fluoroacetate, Sigma-Aldrich®
Ethyl fluoroacetate
Formula: C₄H₇FO₂
MW: 106.1 g/mol
Boiling Pt: 119.2
Density: 1.0912 g/cm³ (20 °C)
MDL Number: MFCD00000450
CAS Number: 459-72-3
EINECS: 207-297-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR