Heptafluorobutyric acid, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
Perfluorobutyric acid, HFBA, Heptafluorobutanoic acid
M-Clarity™ quality level = MQ200
164194-100G
164194-25G
164194-5G
SIAL164194-100GEA
152
EUR
SIAL164194-100G
SIAL164194-25G
SIAL164194-5G
Heptafluorobutyric acid, Sigma-Aldrich®
Heptafluorobutyric acid
Formula:
C₄HF₇O₂ MW: 214.04 g/mol Boiling Pt: 121 Melting Pt: –17.5 °C Density: 1.764 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00004171 CAS Number: 375-22-4 EINECS: 206-786-3 UN: 3265 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...