Heptafluorobutyric acid, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Perfluorobutyric acid, HFBA, Heptafluorobutanoic acid
77249-10ML 77249-50ML
SIAL77249-10MLEA 198 EUR
SIAL77249-10ML SIAL77249-50ML
Heptafluorobutyric acid, Sigma-Aldrich®
Heptafluorobutyric acid
Formula: C₄HF₇O₂
MW: 214.04 g/mol
Boiling Pt: 121
Melting Pt: –17.5 °C
Density: 1.764 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004171
CAS Number: 375-22-4
EINECS: 206-786-3
UN: 3265
ADR: 8 II