Trifluoroacetic anhydride
Supplier: Merck
Danger
Synonyms: Trifluoracetic anhydride, TFAA
106232-500G 106232-25G 106232-100G 106232-10X1G
SIAL106232-500GEA 257 EUR
SIAL106232-500G SIAL106232-25G SIAL106232-100G SIAL106232-10X1G
Trifluoroacetic anhydride
Trifluoroacetic anhydride
Formula: C₄F₆O₃
MW: 210.03 g/mol
Boiling Pt: 39.4
Melting Pt: –65 °C
Density: 1.487 g/cm³ (20 °C)
MDL Number: MFCD00000416
CAS Number: 407-25-0
EINECS: 206-982-9
UN: 3265
ADR: 8 I