Triethylsilyl trifluoromethanesulphonate, Sigma-Aldrich®
Supplier: Merck
Synonyms: Triethylsilyl trifluoromethanesulphonate
M-Clarity™ quality level = MQ100
279471-10G 279471-50G
SIAL279471-10GEA 122 EUR
SIAL279471-10G SIAL279471-50G
Triethylsilyl trifluoromethanesulphonate, Sigma-Aldrich®
Triethylsilyl trifluoromethanesulphonate
Formula: C₇H₁₅F₃O₃SSi
MW: 264.34 g/mol
Boiling Pt: 77…79 °C (1013 hPa)
Density: 1.225 g/cm³ (20 °C)
MDL Number: MFCD00000407
CAS Number: 79271-56-0
EINECS: 279-124-4
UN: 3265
ADR: 8 II