Trimethylsilyl trifluoromethanesulphonate, Sigma-Aldrich®
Supplier: Merck
Synonyms: Trimethylsilyl trifluoromethanesulphonate
M-Clarity™ quality level = MQ200
91741-10ML 91741-250ML 91741-50ML
SIAL91741-10MLEA 65.5 EUR
SIAL91741-10ML SIAL91741-250ML SIAL91741-50ML
Trimethylsilyl trifluoromethanesulphonate, Sigma-Aldrich®
Trimethylsilyl trifluoromethanesulphonate
Formula: C₄H₉F₃O₃SSi
MW: 222.26 g/mol
Boiling Pt: 77
Density: 1.225 g/cm³ (20 °C)
MDL Number: MFCD00000406
CAS Number: 27607-77-8
EINECS: 248-565-4
Merck Index: 12,09849