Trifluoromethanesulphonic acid, Sigma-Aldrich®
Supplier: Merck
Synonyms: Trifluoromethanesulphonic acid
M-Clarity™ quality level = MQ200
158534-10G 158534-50G 158534-100G
SIAL158534-10GEA 37 EUR
SIAL158534-10G SIAL158534-50G SIAL158534-100G
Trifluoromethanesulphonic acid, Sigma-Aldrich®
Trifluoromethanesulphonic acid
Formula: CHF₃O₃S
Boiling Pt: 162 °C (1013 hPa)
Melting Pt: 35 °C
Density: 1.69 g/cm³ (20 °C)
MDL Number: MFCD00007514
CAS Number: 1493-13-6
EINECS: 216-087-5
UN: 3265
ADR: 8,II