1-Iodododecane, Sigma-Aldrich®

Supplier: Merck
Warning


M-Clarity™ quality level = MQ100

238260-100G 238260-25G
SIAL238260-100GEA 135 EUR
SIAL238260-100G SIAL238260-25G
1-Iodododecane, Sigma-Aldrich®
1-Iodododecane
Formula: C₁₂H₂₅I
Boiling Pt: 298 °C (1013 hPa)
Melting Pt: –3 °C
Density: 1.2015 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001088
CAS Number: 4292-19-7
EINECS: 224-293-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR