1-Iodooctane, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ200
238295-100G 238295-25G
SIAL238295-100GEA 129 EUR
SIAL238295-100G SIAL238295-25G
1-Iodooctane, Sigma-Aldrich®
1-Iodooctane
Formula: C₈H₁₇I
Boiling Pt: 225 °C (1013 hPa)
Melting Pt: –46 °C
Density: 1.32 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001105
CAS Number: 629-27-6
EINECS: 211-084-5