Iodoacetonitrile 95%
Supplier: Thermo Fisher Scientific
|
Danger
|
Synonyms:
2-Iodoacetonitrile
122290050
122290250
ACRO122290050EA
105
EUR
ACRO122290050
ACRO122290250
Iodoacetonitrile 95%
Iodoacetonitrile
Formula:
C₂H₂IN MW: 166.95 g/mol Boiling Pt: 182…184 °C (720 mmHg) Density: 2.3 g/cm³ Flash Pt: 86 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00001886 CAS Number: 624-75-9 EINECS: 210-860-0 UN: 3276 ADR: 6.1 II |
Specification Test Results
Appearance | Clear red to brown Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5740 to 1.5780 (20°C, 589 nm) |
GC | ≥94 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...