Iodoacetonitrile, Sigma-Aldrich®
Supplier: Merck
Synonyms: 2-Iodoacetonitrile
M-Clarity™ quality level = MQ200
I6903-10G I6903-50G
SIALI6903-10GEA 106 EUR
SIALI6903-10G SIALI6903-50G
Iodoacetonitrile, Sigma-Aldrich®
Iodoacetonitrile
Formula: C₂H₂IN
Boiling Pt: 182 °C (1013 hPa)
Density: 2.3065 g/cm³ (20 °C)
MDL Number: MFCD00001886
CAS Number: 624-75-9
EINECS: 210-860-0
UN: 3276
ADR: 6.1,III