N,N-Dimethylformamide di-tert-butyl acetal ≥90.0% for GC derivatisation, Sigma-Aldrich®
Supplier: Merck
Synonyms: 1,1-Bis(1,1-dimethylethoxy)-N,N,N-trimethylamine, 1,1-Di-tert-butoxy-N,N-dimethylmethanamine, 1,1-Di-tert-butoxytrimethylamine
M-Clarity™ quality level = MQ100
40263-10ML-F 40263-50ML-F
SIAL40263-10ML-FEA 389 EUR
SIAL40263-10ML-F SIAL40263-50ML-F
N,N-Dimethylformamide di-tert-butyl acetal ≥90.0% for GC derivatisation, Sigma-Aldrich®
N,N-Dimethylformamide di-tert-butyl acetal
Formula: C₁₁H₂₅NO₂
MW: 203.32 g/mol
Boiling Pt: 91…93 °C (1013 hPa)
Density: 0.85 g/cm³ (20 °C)
MDL Number: MFCD00015002
CAS Number: 36805-97-7
EINECS: 253-222-7