N,N-Dimethylformamide diethyl acetal, Sigma-Aldrich®
Supplier: Merck
Synonyms:
1,1-Diethoxy-N,N-dimethylmethanamine, 1,1-Diethoxytrimethylamine
M-Clarity™ quality level = MQ100
142778-10G
142778-50G
SIAL142778-10GEA
60.8
EUR
SIAL142778-10G
SIAL142778-50G
N,N-Dimethylformamide diethyl acetal, Sigma-Aldrich®
N,N-Dimethylformamide diethyl acetal
Formula:
C₇H₁₇NO₂ Boiling Pt: 130 °C (1013 hPa) Density: 0.86 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00009227 CAS Number: 1188-33-6 EINECS: 214-707-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...