Triethanolamine (trolamine)
Supplier: Roth Carl
Synonyms: Tris-(2-hydroxyethyl)amine, 2,2',2''-Nitrilotriethanol
6300.1 6300.2 6300.3
ROTH6300.1EA 0 EUR
ROTH6300.1 ROTH6300.2 ROTH6300.3
Triethanolamine (trolamine)
Triethanolamine (Trolamine)
Formula: C₆H₁₅NO₃
MW: 149.19 g/mol
Boiling Pt: 335.4
Melting Pt: 21.6
Density: 1.13 g/cm³ (20 °C)
Flash Pt: 179 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00002855
CAS Number: 102-71-6
EINECS: 203-049-8
Merck Index: 13,09739
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c181d3e80375aec7c34e17766247484e1c3d3257&vid=40c3d3d0c0162ac22208cadb8d5254a7cf0b340a&prid=web&clid=SAPDAM