2-Diethylaminoethanol 99%
Supplier: Thermo Fisher Scientific
Danger
114330010 114330025 114330100
ACRO114330010EA 31.7 EUR
ACRO114330010 ACRO114330025 ACRO114330100
2-Diethylaminoethanol 99%
2-Diethylaminoethanol
Formula: C₆H₁₅NO
Boiling Pt: 161 °C (760 mmHg)
Melting Pt: –70 °C
Density: 0.88 g/cm³
Flash Pt: 51.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00002850
CAS Number: 100-37-8
EINECS: 202-845-2
UN: 2686
ADR: 8,II
Merck Index: 13,03139
Order Now
Specification Test Results
Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4400 to 1.4420 (20°C, 589 nm)
Water ≤0.5 % (K.F.)
GC ≥98.5 %
Color scale ≤50 APHA