N,N,N',N'-Tetramethylethylenediamine (TEMED), Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ200
T7024-100ML T7024-25ML T7024-50ML
SIALT7024-100MLEA 234 EUR
SIALT7024-100ML SIALT7024-25ML SIALT7024-50ML
N,N,N',N'-Tetramethylethylenediamine (TEMED), Sigma-Aldrich®
N,N,N',N'-Tetramethylethylenediamine (TEMED)
Formula: C₆H₁₆N₂
Boiling Pt: 120 °C (1013 hPa)
Melting Pt: –64 °C
Density: 0.7765 g/cm³ (20 °C)
Flash Pt: 17 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00008335
CAS Number: 110-18-9
EINECS: 203-744-6
UN: 2372
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6838f1f5b7d0f69dcc81a8ed247f6c178dad231c&vid=ea361367e9944e4101e38622fdd18b07756eb676&prid=web&clid=SAPDAM