1,6-Diisocyanatohexane, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Hexamethylene diisocyanate
52649-100ML 52649-25ML
SIAL52649-100MLEA 98 EUR
SIAL52649-100ML SIAL52649-25ML
1,6-Diisocyanatohexane, Sigma-Aldrich®
1,6-Diisocyanatohexane
Formula: C₈H₁₂N₂O₂
MW: 168.2 g/mol
Boiling Pt: 255
Melting Pt: –67 °C
Density: 1.05 g/cm³ (20 °C)
Flash Pt: 130
Storage Temperature: Ambient
MDL Number: MFCD00002047
CAS Number: 822-06-0
EINECS: 212-485-8
UN: 2281
ADR: 6.1 II