1-Butyl isothiocyanate
Supplier: Thermo Scientific
TL00699.1
TL00699.10
TL00699.50
MAYBTL00699.1EA
0
EUR
MAYBTL00699.1
MAYBTL00699.10
MAYBTL00699.50
1-Butyl isothiocyanate
1-Butyl isothiocyanate
Formula:
C₅H₉NS Boiling Pt: 66 °C (1013 hPa) Density: 0,9559 g/cm³ (11.2 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00004824 CAS Number: 592-82-5 EINECS: 209-770-4 UN: 1693 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...