Methyl isothiocyanate, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ200
112771-100G 112771-500G 112771-5G
SIAL112771-100GEA 106 EUR
SIAL112771-100G SIAL112771-500G SIAL112771-5G
Methyl isothiocyanate, Sigma-Aldrich®
Methyl isothiocyanate
Formula: C₂H₃NS
Boiling Pt: 117 °C (1013 hPa)
Melting Pt: 31…37 °C
Density: 1.069 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00004818
CAS Number: 556-61-6
EINECS: 209-132-5
UN: 2477
ADR: 6.1,I
Merck Index: 12,06165