Hexadecylamine, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ200
445312-25G 445312-5G
SIAL445312-25GEA 114 EUR
SIAL445312-25G SIAL445312-5G
Hexadecylamine, Sigma-Aldrich®
Hexadecylamine
Formula: H₃C(CH₂)₁₄CH₂NH₂
Boiling Pt: 330 °C (1013 hPa)
Melting Pt: 43…46 °C
Density: 0.7821 g/cm³ (60 °C)
Flash Pt: 140 °C
Storage Temperature: Ambient
MDL Number: MFCD00008158
CAS Number: 143-27-1
EINECS: 205-596-8
UN: 3259
ADR: 8,II